Learn More
Tashiro's indicator solution, In ethanol, Honeywell Fluka™
Product Code. p-7072189 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
30 mL
250 mL
1 L
1.5 L
6 x 250 mL
6 x 1 L
Unit Size:
1 L
1.5 L
250mL
30mL
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15658390 30 mL 30mL
15645110 250 mL 250mL
15635110 1 L 1 L
15665110 6 x 250 mL Pack of 6
15625110 1.5 L 1.5 L
15655110 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15658390 Supplier Honeywell-Fluka Supplier No. 3608330ML
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
In ethanol
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tashiro’s indicator solution
Quantity 30 mL
Flash Point 12°C (53.6°F)