Learn More
Techne™ Dri-Block™ Heater
Provides a safe, dry, constant temperature source in the laboratory. Dri-Block™ Heaters can be used for incubation, boiling, inactivation, wet ashing, sample concentration, enzyme analysis and many other clinical and industrial purposes.
1201.00€ - 1259.00€
Products 2
Product Code Brand Description Temperature Uniformity Wattage Price Quantity & Availability  
Product Code Brand Description Temperature Uniformity Wattage Price Quantity & Availability  
15285607
View Documents
Techne™
DB100/3
7 minute heat-up time Ambient +5 to 100°C 600 w 1201.00€
Each
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
05-01-2021
Add to basket
15275607
View Documents
Techne™
DB200/3
9 minute heat-up time Ambient +5 to 200°C 450 w 1259.00€
Each
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
05-01-2021
Add to basket
Description
Description
  • The units cover the temperature range from ambient 5°C to 100°C or 200°C (depending on model) using machined aluminium alloy blocks as the heat transfer medium.
  • Independent indicator lights indicate mains power, heater action and over-temperature cut-out. An ON/OFF switch is mounted on the rear panel, together with the mains connector and a double fuse holder.
  • Temperature setting is a simple push button display. Temperatures can be displayed in °C or °F.
  • Built-in count-down timer can be set in 1 minute intervals to 99h 59m.
  • Constructed of painted aluminium alloy and polyphenylene sulphide (PPS) plastic, which is strong yet lightweight and can resist sustained temperatures as high as 260°C.
  • Even though the units heat up rapidly, venting and highly efficient insulation ensure that the cases remain cool enough to handle even at maximum operating temperatures.
  • Each unit is supplied with an extraction tool for easy removal of the insert blocks.
SDS