Learn More
Techne™ Dri-Block™ Heater
Provides a safe, dry, constant temperature source in the laboratory. Dri-Block™ Heaters can be used for incubation, boiling, inactivation, wet ashing, sample concentration, enzyme analysis and many other clinical and industrial purposes.
1201.00€ - 1259.00€
| Product Code | Brand | Description | Temperature Uniformity | Wattage | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Description | Temperature Uniformity | Wattage | Price | Quantity & Availability | |||
|
15285607
|
Techne™
DB100/3 |
7 minute heat-up time | Ambient +5 to 100°C | 600 w |
1201.00€
Each Qty Price Break Available |
05-01-2021 |
15275607
|
Techne™
DB200/3 |
9 minute heat-up time | Ambient +5 to 200°C | 450 w |
1259.00€
Each Qty Price Break Available |
05-01-2021 |
Description
- The units cover the temperature range from ambient 5°C to 100°C or 200°C (depending on model) using machined aluminium alloy blocks as the heat transfer medium.
- Independent indicator lights indicate mains power, heater action and over-temperature cut-out. An ON/OFF switch is mounted on the rear panel, together with the mains connector and a double fuse holder.
- Temperature setting is a simple push button display. Temperatures can be displayed in °C or °F.
- Built-in count-down timer can be set in 1 minute intervals to 99h 59m.
- Constructed of painted aluminium alloy and polyphenylene sulphide (PPS) plastic, which is strong yet lightweight and can resist sustained temperatures as high as 260°C.
- Even though the units heat up rapidly, venting and highly efficient insulation ensure that the cases remain cool enough to handle even at maximum operating temperatures.
- Each unit is supplied with an extraction tool for easy removal of the insert blocks.