Learn More
Techne™ 5Prime Thermal Cyclers: Prime Gradient Upgrade
Product Code. 12612956
Change view
Click to view available options
:
Prime gradient upgrade, 100-230V 50-60Hz, 450w
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12612956 Prime gradient upgrade, 100-230V 50-60Hz, 450w
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12612956 Supplier Techne™ Supplier No. 9394557
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This is an UPGRADE only. Upgrade the base model of Techne 5Prime Thermal Cycler to gradient capability.
Prime units can be upgraded to achieve gradient capability. The gradient function can be enabled at any time without the need to change a block. The simple process is installed via an unlock code.
Specifications
Product Type Prime Gradient Upgrade