missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagJET™ Whole Blood RNA Kit

Uses magnetic bead technology to purify total RNA from fresh or frozen whole blood treated with EDTA or citrate, blood fractions, bone marrow, or samples stabilized with PAXgene™ or Tempus™.

Brand:  Thermo Scientific™ K2751

Additional Details : Weight : 0.25000kg Transport : UN number : 1502 Chem class : 5.1 Pack group : II

Product Code. 13535180

  • 376.00€ / 96 tests


Estimated Shipment date
from Supplier 24-06-2020
Add to basket

Description

Description

The Thermo Scientific MagJET Whole Blood RNA Kit quickly and efficiently purifies total RNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions and bone marrow samples. The kit is also suitable for total RNA purification from human blood samples stabilized with commercial stabilizers such as PAXgene™ or Tempus™.
Paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual nucleic acid purification by low sample throughput users.
The resulting high quality purified RNA is free of proteins, nucleases and other contaminants or inhibitors, and can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR and other enzymatic reactions.

Highlights

  • Versatile - suitable for total RNA purification from fresh, stabilized, frozen or aged blood, as well as bone marrow or buffy coat
  • Efficient - high yields of high-integrity RNA
  • High quality - isolates intact total RNA, free of inhibitors and gDNA contamination
  • Flexible - easily adjustable for high throughput automation platforms and manual processing
Applications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
  • RT-PCR
  • RT-qPCR
  • Northern blotting
  • Nuclease protection assay
Includes
  • Lysis Buffer
  • Proteinase K
  • MagJET Magnetic Beads
  • DNase I (lyophilized)
  • 2X DNase I Buffer
  • Manganese Chloride Solution
  • DNase I Reconstitution Buffer
  • Rebinding Buffer
  • Wash Buffer 1 (conc.)
  • Wash Buffer 2 (conc.)
  • Water, nuclease free

Hazardous: Yes
Hazardous: No
Quality Control: The MagJET Whole Blood RNA Kit is qualified by isolating total RNA from 250 μL of fresh human blood following the protocols outlined in the manual. The purified total RNA has an A260/A280 ratio between 1.9 and 2.1. The quality of isolated RNA is evaluated spectrophotometrically and by agarose gel electrophoresis. The functional quality of purified RNA is evaluated by RT-qPCR.
Shelf Life:24 Months
Shipping Condition: Ambient
Storage Condition: Ambient

Specifications

Specifications

High purity RNA suitable for all conventional molecular biology procedures like RT-PCR, RT-qPCR, Northern blotting, Nuclease protection assay
Lysis Buffer, Proteinase K, MagJET Magnetic Beads, DNase I (lyophilized), 2X DNase I Buffer, Manganese Chloride Solution, DNase I Reconstitution Buffer, Rebinding Buffer, Wash Buffer 1 (conc.), Wash Buffer 2 (conc.), Water, nuclease free
96 Preparations
DNase I (lyophilized), 2X DNase I Buffer, DNase I Reconstitution Buffer and reconstituted DNase I solution should be stored at -20deg.C. Proteinase K solution is stable at room temperature as long as the vial remains sealed. After being opened, Proteinase K solution should be stored at −20deg.C. MagJET Magnetic Beads should be stored at 4deg.C. Other components of the kit should be stored at room temperature (15deg. to 25deg.C).
Videos
Documents