Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Rectangular Buckets
Product Code. 10140941 Shop All Thermo Scientific Products
Change view
Click to view available options
Tube Type:
15 mL Blood Collection Tube
15 mL DIN
Conical
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Type unitSize
10140941 15 mL DIN Each
10216842 15 mL Blood Collection Tube Each
10059430 Conical Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10140941 Supplier Thermo Scientific™ Supplier No. 75005322
Only null left
Add to basket
Add to basket
For use with four-place swinging bucket rotor 75-002-000 when equipped with rectangular bucket 75-002-001
Adapters sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
Tube Type 15 mL DIN
For Use With (Equipment) 4-place Swing rotor with rectangle bucket