missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Trichloroacetic acid, 5% w/v aq. soln.
Protein precipitation reagent
64.90€ - 94.33€
Chemical Identifiers
| CAS | 76-03-9 |
|---|---|
| Molecular Formula | C2HCl3O2 |
| Molecular Weight (g/mol) | 163.378 |
| MDL Number | MFCD00004177 |
| InChI Key | YNJBWRMUSHSURL-UHFFFAOYSA-N |
| Synonym | trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer |
| PubChem CID | 6421 |
| ChEBI | CHEBI:30956 |
| IUPAC Name | 2,2,2-trichloroacetic acid |
| SMILES | C(=O)(C(Cl)(Cl)Cl)O |
Description
ApplicationsTrichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.
Solubility
Soluble in water (1200 mg/ml at 20°C), alcohol, ether, ethanol (very soluble), and carbon tetrachloride.
Notes
Store under nitrogen. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases, smines.
Chemical Identifiers
| 76-03-9 | |
| 163.378 | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 6421 | |
| 2,2,2-trichloroacetic acid |
| C2HCl3O2 | |
| MFCD00004177 | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| CHEBI:30956 | |
| C(=O)(C(Cl)(Cl)Cl)O |
Specifications
| Trichloroacetic acid | |
| C2HCl3O2 | |
| MFCD00004177 | |
| 970119 | |
| trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
| YNJBWRMUSHSURL-UHFFFAOYSA-N | |
| 2,2,2-trichloroacetic acid | |
| 5% w/v aq. soln. | |
| CHEBI:30956 | |
| Liquid | |
| Colorless |
| 76-03-9 | |
| Cl3CCO2H | |
| UN2564 | |
| 14,9627 | |
| Soluble in water (1200mg/ml at 20°C),alcohol,ether,ethanol (very soluble),and carbon tetrachloride. | |
| C(=O)(C(Cl)(Cl)Cl)O | |
| 163.378 | |
| 6421 | |
| 163.39 | |
| Acrid | |
| 500mL |