Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tris(triphenylphosphine)rhodium(I) chloride, 99.99%, (trace metal basis), ACROS Organics™
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
Description
For ultra trace analysis
Specifications
| Quantity | 250mg |
| Packaging | Glass bottle |
| Infrared Spectrum | Authentic |
| Melting Point | 245.0°C to 250.0°C |
| Color | Maroon |
| Assay Percent Range | 99.99% |
| Linear Formula | [(C6H5)3P]3RhCl |
| Fieser | 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,90 |
| Merck Index | 15, 10245 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in chloroform, dichloromethane, benzene,, toluene, less soluble in acetic acid, ketones, methanol and, lower aliphatic alcohols, virtually insoluble in alkanes and cyclohexane |
| Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Warning
EINECSNumber : 238-744-5
TSCA : TSCA