Learn More
Thermo Scientific™ TX-750 Round Bucket Adapters
Product Code. 11864461 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
1.5/2 mL Conical Microtube
100 mL Round-bottom Open-top Tube
14 mL Conical Urine Tube
15 mL Conical Tube
25 mL Universal Conical/Skirted Tube
250 mL Bottle
250 mL Corning Conical Bottle
5 mL RIA/Round-bottom Tube (w/o cap)
5/7 mL Blood Collection Tube
5/7 mL Round-bottom Tube (w/o cap) w/Decanting Aid
50 mL Conical Tube
50 mL Round-bottom Tube
84 x 15 mL Blood Collection Tube
Unit Size:
Pack of 4
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
11864461 14 mL Conical Urine Tube Pack of 4
10491445 50 mL Conical Tube Pack of 4
10287707 15 mL Conical Tube Pack of 4
11894451 250 mL Bottle Pack of 4
10595494 100 mL Round-bottom Open-top Tube Pack of 4
11824461 50 mL Round-bottom Tube Pack of 4
11844461 25 mL Universal Conical/Skirted Tube Pack of 4
11874461 84 x 15 mL Blood Collection Tube Pack of 4
11884461 5/7 mL Blood Collection Tube Pack of 4
11894461 5 mL RIA/Round-bottom Tube (w/o cap) Pack of 4
11804471 5/7 mL Round-bottom Tube (w/o cap) w/Decanting Aid Pack of 4
11814471 1.5/2 mL Conical Microtube Pack of 4
11953759 250 mL Corning Conical Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11864461 Lieferant Thermo Scientific™ Lieferanten-Nr. 75003718
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
TRUSTED_SUSTAINABILITY
Spezifikation
Type Round Bucket Adapter
Tube Type 14 mL Conical Urine Tube
Capacity 14 mL Conical Urine Tube
For Use With (Equipment) Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube