Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Small Bag
Designed for the steam sterilization of large or odd shaped pharmaceutical equipment change parts. These bags are closed via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically opened without having to cut or remove the particulate generating autoclave tape.
1480.00€ - 3550.00€
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | COC |
Closure Type | Drawstring |
Color | White |
Disposable | Single-use |
Product Code | Brand | Capacity (English) | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Dimensions (L x W) | Price | Quantity & Availability | ||||
17747887
|
Keystone Cleanroom Products
BGT2A0403D |
9 oz. | 4 x 3 in. |
3095.00€
Pack of 500 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17757887
|
Keystone Cleanroom Products
BGT2A0404D |
12 oz. | 4 x 4 in. |
3130.00€
Pack of 500 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17787887
|
Keystone Cleanroom Products
BGT2A0410D |
30 oz. | 10 x 4 in. |
3550.00€
Pack of 500 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17707897
|
Keystone Cleanroom Products
BGT2A0418D |
54 oz. | 18 x 4 in. |
2080.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17717897
|
Keystone Cleanroom Products
BGT2A0425D |
75 oz. | 25 x 4 in. |
2305.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17727897
|
Keystone Cleanroom Products
BGT2A0426D |
78 oz. | 26 x 4 in. |
2340.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17777897
|
Keystone Cleanroom Products
BGT2A0505D18 |
20 oz. | 5 x 5 in. |
2010.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17787897
|
Keystone Cleanroom Products
BGT2A0506D |
24 oz. | 6 x 5 in. |
1850.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17797897
|
Keystone Cleanroom Products
BGT2A0508D |
32 oz. | 8 x 5 in. |
1875.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17707907
|
Keystone Cleanroom Products
BGT2A0509D |
36 oz. | 9 x 5 in. |
1930.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17717907
|
Keystone Cleanroom Products
BGT2A0510D |
40 oz. | 10 x 5 in. |
1960.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17727907
|
Keystone Cleanroom Products
BGT2A0511D |
44 oz. | 11 x 5 in. |
1995.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17757907
|
Keystone Cleanroom Products
BGT2A0606D |
30 oz. | 6 x 6 in. |
1480.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17767907
|
Keystone Cleanroom Products
BGT2A0608D |
40 oz. | 8 x 6 in. |
1580.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17787907
|
Keystone Cleanroom Products
BGT2A0609D |
45 oz. | 9 x 6 in. |
1645.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17797907
|
Keystone Cleanroom Products
BGT2A0610D |
50 oz. | 10 x 6 in. |
1650.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17727917
|
Keystone Cleanroom Products
BGT2A0615D |
75 oz. | 15 x 6 in. |
2170.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17747917
|
Keystone Cleanroom Products
BGT2A0616D |
80 oz. | 16 x 6 in. |
2190.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17787917
|
Keystone Cleanroom Products
BGT2A0707D |
42 oz. | 7 x 7 in. |
1700.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17707927
|
Keystone Cleanroom Products
BGT2A0709D |
54 oz. | 9 x 7 in. |
1740.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17727927
|
Keystone Cleanroom Products
BGT2A0710D |
60 oz. | 10 x 7 in. |
1790.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17767927
|
Keystone Cleanroom Products
BGT2A0811D |
77 oz. | 11 x 8 in. |
2320.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
17797997
|
Keystone Cleanroom Products
BGT2A1420D |
20 x 14 in. |
2955.00€
Pack of 250 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
Description
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Eliminates need to cut material and the use of autoclave tape
- Fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Sealed seams eliminate sewing threads and greatly reduce particulates
- Resists tearing and punctures
- Superior microbial barrier
- Lint-free Category 1 (Helmke Drum)
- Manufactured and packaged in a ISO 6 cleanroom environment
- Certificates of Conformance sent with shipment
Specifications
Autoclavable | |
Drawstring | |
Single-use | |
Pharmaceutical Equipment | |
Non-sterile | |
<250°F |
COC | |
White | |
Steam Sterilization | |
Tyvek | |
Steam | |
<125°C |