Learn More
UVP UV lamp UVL-24 EL hand held 220V 50Hz 4W 254nm UVP
Brand:  UVP 95026902
Additional Details : Weight : 0.80000kg
Product Code. 11788251
Quantity Price
1 355.00€ / Each
6 305.44€ / Each
Estimated Shipment
17-02-2021
Add to basket