missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Waterproof Portable pH Meter

Product Code. 11795274
Change view
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
11795274 -2 to 16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11795274 Supplier Hanna Instruments™ Supplier No. HI9124N

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Designed to provide laboratory precision under harsh environmental and industrial conditions. Hanna Instruments™ Waterproof Portable pH Meter is a rugged meter that offers many features and is supplied complete with meter, pH electrode, temperature probe and a plastic carrying case.

This meter offers a variety of features including automatic calibration to 1 or 2 points, automatic temperature compensation, backlit LCD, on-screen instructions, and battery error prevention system. It uses the universal BNC probe input to allows the user to select from a wide variety of electrodes based on their particular application. A separate RCA connector is used for a thermistor temperature probe. The temperature probe provides for a high ±0.4°C accuracy measurement for precise temperature compensation.

  • Waterproof to IP67 standards
  • Automatic Calibration: At 1 or 2 points using pre-programmed buffers (pH 4.01, 6.86, 7.01, 9.18, and 10.01)
  • On-Screen Instructions: Clear and easy to use steps for calibration and taking measurements
  • Durable Plastic Body pH Electrode: Supplied with the HI1230B PEI plastic body, double junction pH electrode that is gel filled
  • Temperature Compensation: Automatic or manual temperature compensation of pH measurements -20.0 to 120.0°C
  • Measurement Recall: Press 'MEM' to store a reading, recall it at anytime by pressing the 'MR' button
  • Automatic Shut off: After 20 minutes, to prevent accidental drainage of the batteries (it can be disabled)
  • Battery Error Prevention System (BEPS): Detects when the batteries are too weak to ensure reliable measurements

Specifications

Includes HI1230B pH electrode, HI7662 temperature probe, 1m cable, HI70004 pH 4.01 buffer solution sachet, HI70007 pH 7.01 buffer sachet, 100mL plastic beaker, 1.5V AAA batteries (3), instructions, and hard carrying case
Accuracy (Temperature) ±0.4°C/±0.8°F
Accuracy (pH) ±0.01
Calibration Automatic
Range (pH) -2 to 16
Resolution (pH) 0.01
Calibration Points 1 or 2
Temperature Range (Metric) -20.0°C to +120.0°C
Temperature Range (English) -4.0°F to +248.0°F
Resolution (Temperature) 0.1°C, 1°F
Display Type LCD, Backlit
Backlight Option Yes
Alarm Output Low battery
Certifications/Compliance IP67
Dimensions (L x W x H) 185 x 72 x 36 mm (7.3 x 2.8 x 1.4 in.)
Weight (Metric) 300 g
Weight (English) 10.6 oz.
Warranty 2 Years (6 Months Probe)
Material Temp probe: stainless steel, pH electrode: PEI body
Probe Type HI1230B PEI body pH electrode with BNC connector, HI7662 stainless steel temperature probe
Relative Humidity Range 100% max.
Temperature Compensation Automatic or manual, -20 to 120°C
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Operating 0°C to 50°C
Battery Type 3 x 1.5V AAA
Battery Life Auto-off after 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.