Learn More
Fisherbrand™ Grade 908 Weighing Papers
Product Code. p-8003198 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
150 x 150mm (5.9 x 5.9 in.)
95 x 110mm (3.7 x 4.3 in.)
Unit Size:
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
11754576 150 x 150mm (5.9 x 5.9 in.) Pack of 250
11744576 95 x 110mm (3.7 x 4.3 in.) Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11754576
Add to basket
Edge
Add to basket
Smooth paper with satin appearance and wetstrength resistance.
  • Smooth paper 45g/m2
  • With satin appearance, wetstrength resistance
Specifications
For Use With (Application) Weighing of all types of substances such as beet mash, powders and liquids
Dimensions (L x W) 150 x 150mm (5.9 x 5.9 in.)
Length (English) 5.9 in.
Length (Metric) 150 mm
Material Paper
Width (English) 5.9 in.
Width (Metric) 150 mm
Product Type Weighing Paper
Shape Square