accessibility menu, dialog, popup

Test

CAS RN 100-40-3

CAS RN 100-40-3

IUPAC Name: 4-ethenylcyclohex-1-ene
Synonyms: 4-vinylcyclohexene
Molecular Weight (g/mol): 108.18
Molecular Formula: C8H12
InChi Key: BBDKZWKEPDTENS-UHFFFAOYNA-N
SMILES: C=CC1CCC=CC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)