accessibility menu, dialog, popup

Test

CAS RN 1000018-58-5

CAS RN 1000018-58-5

IUPAC Name: 3,5-dibromo-2-chloro-6-methylpyridine
Synonyms: 3,5-dibromo-2-chloro-6-methylpyridine
Molecular Weight (g/mol): 285.36
Molecular Formula: C6H4Br2ClN
InChi Key: AYZAQZJMQIXSLF-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results