accessibility menu, dialog, popup

Test

CAS RN 10025-83-9

ClClClIr

CAS RN 10025-83-9

IUPAC Name: iridium(3+) trichloride
Synonyms: iridium(III) chloride
Molecular Weight (g/mol): 298.57
Molecular Formula: Cl3Ir
InChi Key: DANYXEHCMQHDNX-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Ir+3]
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results