accessibility menu, dialog, popup

Test

CAS RN 10025-94-2

H2OH2OH2OH2OH2OH2OClClClY

CAS RN 10025-94-2

IUPAC Name: yttrium(3+) hexahydrate trichloride
Synonyms: yttrium(III) chloride
Molecular Weight (g/mol): 303.35
Molecular Formula: Cl3H12O6Y
InChi Key: IINACGXCEZNYTF-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Y+3]
Grade 
  • (3)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)