accessibility menu, dialog, popup

Test

CAS RN 10025-99-7

ClPtClKKClCl

CAS RN 10025-99-7

IUPAC Name: dipotassium tetrachloroplatinumdiuide
Synonyms: potassium tetrachloroplatinate
Molecular Weight (g/mol): 419.11
Molecular Formula: Cl4H4K2Pt
InChi Key: JCAXKWJYKUEOFO-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]
Physical Form 
  • (5)
  • (3)
  • (2)
  • (1)
Quantity 
  • (5)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (4)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (5)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (2)