accessibility menu, dialog, popup

Test

CAS RN 1002727-87-8

NCOO

CAS RN 1002727-87-8

IUPAC Name: 6-isocyanato-3,4-dihydro-2H-1-benzopyran
Synonyms: 6-isocyanato-3,4-dihydro-2H-1-benzopyran
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: YNAWMZLOWDUDQF-UHFFFAOYSA-N
SMILES: O=C=NC1=CC2=C(OCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results