accessibility menu, dialog, popup

Test

CAS RN 1006-33-3

H3COFBr

CAS RN 1006-33-3

IUPAC Name: 1-(2-bromo-5-fluorophenyl)ethan-1-one
Synonyms: 1-(2-bromo-5-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: BQXROZQUDCJBBY-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(Br)C=CC(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results