CAS RN 1006028-37-0
CAS RN 1006028-37-0
IUPAC Name:
{[2-(3-{3-[({3-[(5-{[2,4-bis(phosphonomethyl)phenyl]carbamoyl}-2-methylphenyl)carbamoyl]phenyl}carbamoyl)amino]benzamido}-4-methylbenzamido)-5-(phosphonomethyl)phenyl]methyl}phosphonic acid tetrasodium
Synonyms:
[2-(3-{3-[({3-[(5-{[2,4-bis(phosphonomethyl)phenyl]carbamoyl}-2-methylphenyl)carbamoyl]phenyl}carbamoyl)amino]benzamido}-4-methylbenzamido)-5-(phosphonomethyl)phenyl]methylphosphonic acid tetrasodium
Molecular Weight (g/mol):
1184.78
Molecular Formula:
C47H48N6Na4O17P4
InChi Key:
HLUPMJLBMAPXSC-UHFFFAOYSA-N
SMILES:
[Na].[Na].[Na].[Na].CC1=C(NC(=O)C2=CC(NC(=O)NC3=CC=CC(=C3)C(=O)NC3=C(C)C=CC(=C3)C(=O)NC3=C(CP(O)(O)=O)C=C(CP(O)(O)=O)C=C3)=CC=C2)C=C(C=C1)C(=O)NC1=C(CP(O)(O)=O)C=C(CP(O)(O)=O)C=C1