accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10075-62-4

CH3OOH3C

CAS RN 10075-62-4

IUPAC Name: 1,4-dimethoxynaphthalene
Synonyms: 1,4-dimethoxynaphthalene
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: FWWRTYBQQDXLDD-UHFFFAOYSA-N
SMILES: COC1=C2C=CC=CC2=C(OC)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.