accessibility menu, dialog, popup

Test

CAS RN 100888-40-2

H3CBOHOH

CAS RN 100888-40-2

IUPAC Name: tetradecylboronic acid
Synonyms: tetradecylboronic acid
Molecular Weight (g/mol): 242.21
Molecular Formula: C14H31BO2
InChi Key: MCUAMUGSKSODIJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCB(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results