CAS RN 100960-07-4
CAS RN 100960-07-4
IUPAC Name:
1H-pyrrolo[2,3-b]pyridin-5-amine
Synonyms:
1H-pyrrolo[2,3-b]pyridin-5-amine
Molecular Weight (g/mol):
133.15
Molecular Formula:
C7H7N3
InChi Key:
PLWBENCHEUFMTN-UHFFFAOYSA-N
SMILES:
NC1=CN=C2NC=CC2=C1
1
–
1
of
1
results
1
5-Amino-7-azaindole, 97%, Thermo Scientific™
CAS: 100960-07-4 Molecular Formula: C7H7N3 Molecular Weight (g/mol): 133.154 MDL Number: MFCD06659683 InChI Key: PLWBENCHEUFMTN-UHFFFAOYSA-N Synonym: 5-amino-7-azaindole,1h-pyrrolo 2,3-b pyridin-5-amine,1h-pyrrolo 2,3-b pyridin-5-ylamine,1h-pyrrolo 2,3-b pyridine-5-amine,5-amino-1h-pyrrolo 2,3-b pyridine,5-amino-7-aza-1h-indole,pubchem15250,acmc-2097tg,pyrrolo 2,3-b pyrid-5-ylamine,pyrrolo 2,3-b pyridin-5-yl amine PubChem CID: 11715187 IUPAC Name: 1H-pyrrolo[2,3-b]pyridin-5-amine SMILES: C1=CNC2=NC=C(C=C21)N