accessibility menu, dialog, popup

Test

CAS RN 101-01-9

CAS RN 101-01-9

IUPAC Name: N,N',N''-triphenylguanidine
Synonyms: guanidine, N,N',N''-triphenyl-
Molecular Weight (g/mol): 287.37
Molecular Formula: C19H17N3
InChi Key: FUPAJKKAHDLPAZ-UHFFFAOYSA-N
SMILES: N(C(NC1=CC=CC=C1)=NC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results