man

Test

CAS RN 101-68-8

OCNNCO

CAS RN 101-68-8

IUPAC Name: 1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
Synonyms: diphenylmethane diisocyanate methylbisphenyl isocyanate MDI MDR diphenyl methane diisocyanate methylene diphenyl diisocyanate diphenylmethyl diisocyanate methylene bisphenyl isocyanate
Molecular Weight (g/mol): 250.26
Molecular Formula: C15H10N2O2
InChi Key: UPMLOUAZCHDJJD-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=C(CC2=CC=C(C=C2)N=C=O)C=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)