accessibility menu, dialog, popup

Test

CAS RN 10102-23-5

CAS RN 10102-23-5

IUPAC Name: disodium decahydrate selenate
Synonyms: disodium decahydrate selenate
Molecular Weight (g/mol): 369.1
Molecular Formula: H20Na2O14Se
InChi Key: TYVNFQRMDPFYPC-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results