accessibility menu, dialog, popup

Test

CAS RN 10102-25-7

LiLiH2OOSOOO

CAS RN 10102-25-7

IUPAC Name: dilithium(1+) hydrate sulfate
Synonyms: dilithium(1+) hydrate sulfate
Molecular Weight (g/mol): 127.95
Molecular Formula: H2Li2O5S
InChi Key: RKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES: [Li+].[Li+].O.[O-]S([O-])(=O)=O
Quantity 
  • (2)
  • (8)
  • (1)
  • (1)
  • (7)
  • (1)
Percent Purity 
  • (1)
  • (4)
  • (2)
  • (2)
  • (5)
Grade 
  • (3)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (1)
  • (6)
  • (6)
  • (1)
  • (2)
  • (8)
  • (1)
  • (1)
  • (7)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)