accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1011-54-7

CH3OHOO

CAS RN 1011-54-7

IUPAC Name: (2E)-3-(2-methoxyphenyl)prop-2-enoic acid
Synonyms: (2E)-3-(2-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: FEGVSPGUHMGGBO-VOTSOKGWSA-N
SMILES: COC1=CC=CC=C1\C=C\C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.