accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103639-04-9

CAS RN 103639-04-9

IUPAC Name: hydrogen 9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-2,3,4,9-tetrahydro-1H-carbazol-4-one dihydrate chloride
Synonyms: hydrogen ondansetron dihydrate chloride
Molecular Weight (g/mol): 365.86
Molecular Formula: C18H24ClN3O3
InChi Key: VRSLTNZJOUZKLX-UHFFFAOYNA-N
SMILES: [H+].O.O.[Cl-].CN1C2=C(C3=CC=CC=C13)C(=O)C(CN1C=CN=C1C)CC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.