accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1072944-24-1

CAS RN 1072944-24-1

IUPAC Name: [4-formyl-3-(trifluoromethyl)phenyl]boronic acid
Synonyms: 4-formyl-3-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 217.94
Molecular Formula: C8H6BF3O3
InChi Key: OQDHNEKEJVPVDS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=C(C=O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.