accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 886851-48-5

CAS RN 886851-48-5

IUPAC Name: methyl({[4-(pyrimidin-2-yl)phenyl]methyl})amine
Synonyms: methyl({[4-(pyrimidin-2-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 199.26
Molecular Formula: C12H13N3
InChi Key: DIWVUAYHEYROLA-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.