missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oct-3/4 Antibody (240408), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1759T
This item is not returnable.
View return policy
Description
Oct-3/4 Monoclonal antibody specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
Oct-3/4 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5460 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
240408 | |
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
OCT3,Oct-3,OCT4,Oct-4,OCT4,Oct3/4,OTF3,OTF-3,OTF4,POU domain protein,POU domain, class 5, transcription factor 1,POU5F1 | |
E. coli-derived recombinant human Oct-3/4, Met1-Asn265 (Met262Leu), Accession # Q01860 | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oct-3/4 Antibody (240408), Alexa Fluor™ 594, Novus Biologicals™ > 0.1 mL, Alexa Fluor 594
Spot an opportunity for improvement?Share a Content Correction