Learn More
Axygen™ 0.2 mL Polypropylene PCR Tube Strips
Available in a variety of colors for easy sample identification. Axygen™ 8-Strip PCR Tubes, 0.2 mL are constructed of sturdy polypropylene. Each non-sterile, conical tube features thin and consistent wall thickness. Sold with flat or domed caps to prevent sample loss or without caps.
1420.84€
Specifications
Color Clear
Volume (Metric) 0.2 mL
Sterility Non-sterile
View More Specs
Products 2
Product Code Brand Closure Type Price Quantity & Availability  
Product Code Brand Closure Type Price Quantity & Availability  
16272591
View Documents
Axygen™
PCR-0208-AD-C
Domed Cap
1420.84€
Pack of 1260
16282591
View Documents
Axygen™
PCR-0208-AF-C
Flat Cap
1420.84€
Pack of 1260
Description
Description
Axygen™ 0.2 mL thin-wall 8 strip PCR tubes, available in an assortment of colors are designed to fit a wide variety of thermal cyclers. Ultra-thin and consistent wall thickness allows precise thermal transfer, even insertion cap ensures a tight fit to prevent evaporation.
  • RNase-/DNase-free
  • Nonpyrogenic
  • non-sterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Specifications
    Specifications
    Clear
    0.2 mL
    Non-sterile
    Documents
    Product Certifications
    Product Content Correction
    Product Title
    Axygen™ 0.2 mL Polypropylene PCR Tube Strips