Learn More
1,1,4,4-Tetraphenyl-1,3-butadiene, 99%, Thermo Scientific Chemicals
43.11€ - 132.66€
Chemical Identifiers
CAS | 1450-63-1 |
---|---|
Molecular Formula | C28H22 |
Molecular Weight (g/mol) | 358.48 |
MDL Number | MFCD00004766 |
InChI Key | KLCLIOISYBHYDZ-UHFFFAOYSA-N |
Synonym | 1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene |
PubChem CID | 74060 |
SMILES | C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493447
|
Thermo Scientific Alfa Aesar
A17225.03 |
1 g |
43.11€
1g |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
11403457
|
Thermo Scientific Alfa Aesar
A17225.06 |
5 g |
132.66€
5g |
In Stock
Log in to see stock. |
|||||
Description
1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.
Solubility
Soluble in ethanol, benzene, chloroform, tolune and acetic acid.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
1450-63-1 | |
358.48 | |
KLCLIOISYBHYDZ-UHFFFAOYSA-N | |
74060 |
C28H22 | |
MFCD00004766 | |
1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene | |
C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
1450-63-1 | |
C28H22 | |
1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene | |
KLCLIOISYBHYDZ-UHFFFAOYSA-N | |
1,4,4-triphenylbuta-1,3-dienylbenzene | |
74060 | |
99% |
197°C to 202°C | |
MFCD00004766 | |
Soluble in ethanol,benzene,chloroform,tolune and acetic acid. | |
C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1 | |
358.48 | |
358.48 | |
1,1,4,4-Tetraphenyl-1,3-butadiene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 215-914-7
missing translation for 'rtecsNumber' : CY9040630
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.