missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Phenanthroline Monohydrochloride Monohydrate, 97%, Thermo Scientific Chemicals
52.96€ - 310.55€
Chemical Identifiers
CAS | 18851-33-7 |
---|---|
Molecular Formula | HCl·H2O |
Molecular Weight (g/mol) | 234.69 |
MDL Number | MFCD00150061 |
InChI Key | NDLHUHRGAIHALB-UHFFFAOYSA-N |
Synonym | 1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309 |
PubChem CID | 2723715 |
IUPAC Name | 1,10-phenanthroline;hydrate;hydrochloride |
SMILES | C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011480
|
Thermo Scientific Acros
130140100 |
10 g | Glass bottle |
52.96€
10g |
In Stock
Log in to see stock. |
||||
10294840
|
Thermo Scientific Acros
130140250 |
25 g | Glass bottle |
94.27€
25g |
In Stock
Log in to see stock. |
||||
10629263
|
Thermo Scientific Acros
130141000 |
100 g | Glass bottle |
310.55€
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18851-33-7 | |
234.69 | |
NDLHUHRGAIHALB-UHFFFAOYSA-N | |
2723715 | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl |
HCl·H2O | |
MFCD00150061 | |
1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309 | |
1,10-phenanthroline;hydrate;hydrochloride |
Specifications
18851-33-7 | |
100.0 | |
Purple or White to Yellow | |
97% | |
MFCD00150061 | |
Solubility in water: freely soluble. | |
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl | |
234.69 | |
234.69 | |
Powder |
96.0 | |
224.0°C to 225.0°C | |
Authentic | |
HCl·H2O | |
1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309 | |
NDLHUHRGAIHALB-UHFFFAOYSA-N | |
1,10-phenanthroline;hydrate;hydrochloride | |
2723715 | |
97% | |
1, 10-Phenanthroline monohydrochloride monohydrate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1,10-Phenanthroline monohydrochloride monohydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only