missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dibromo-2-chloro-5-fluorobenzene, 98%, Thermo Scientific Chemicals Product Code.: 11427056

1,3-Dibromo-2-chloro-5-fluorobenzene, 98%, Thermo Scientific Chemicals

Product Code. 11427056
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11427056

Brand: Thermo Scientific Alfa Aesar B25376.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179897-90-6
Molecular Formula C6H2Br2ClF
Molecular Weight (g/mol) 288.34
MDL Number MFCD00061122
InChI Key PZKDJJMHRYNBOR-UHFFFAOYSA-N
Synonym 1-chloro-2,6-dibromo-4-fluorobenzene, 2,6-dibromo-4-fluorochlorobenzene, 2-chloro-1,3-dibromo-5-fluorobenzene, 4-chloro-3,5-dibromo-1-fluorobenzene, benzene, 1,3-dibromo-2-chloro-5-fluoro, pubchem3411, acmc-1c9lw, 2,6-dibromo-4-fluoro-1-chlorobenzene, benzene,1,3-dibromo-2-chloro-5-fluoro
PubChem CID 2724604
IUPAC Name 1,3-dibromo-2-chloro-5-fluorobenzene
SMILES FC1=CC(Br)=C(Cl)C(Br)=C1

Specifications

Melting Point 84°C to 87°C
Quantity 5 g
Formula Weight 288.35
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-2-chloro-5-fluorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dibromo-2-chloro-5-fluorobenzene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.