missing translation for 'onlineSavingsMsg'
Learn More
1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99% Product Code.: 10205800

1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%

Product Code. 10205800 Shop All Thermo Scientific Chemicals Products
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10205800

Brand: Thermo Scientific Acros 170690500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72934-37-3 | C10H9ClO2 | 196.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72934-37-3
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD00001289
InChI Key YAHLWSGIQJATGG-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopropanecarboxylic acid, 1-4-chlorophenyl cyclopropane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-4-chlorophenyl, 1-p-chlorophenyl cyclopropanecarboxylic acid, acmc-209oos, 1-4-chlorophenyl cyclopropanecarboxylicacid, 1-4chloro-phenyl-cyclopropylcarboxylic acid, 1-4-chloro-phenyl-cyclopropylcarboxylic acidShow MoreShow Less
PubChem CID 98606
IUPAC Name 1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
SMILES C1CC1(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 152.0°C to 155.0°C
CAS Min % 98.5
Color White
CAS Max % 100.0
Quantity 50 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 99%
Linear Formula ClC6H4C3H4CO2H
Formula Weight 196.63
Percent Purity 99%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0430506 Certificate TypeCertificate of Analysis Date27/11/2023 Catalog Number170690500,170690100
Lot NumberA0372111 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number170690500,170690100
Lot NumberA0425551 Certificate TypeCertificate of Analysis Date21/12/2020 Catalog Number170690500,170690100
compliance-icons
Product Identifier
  • 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.