missing translation for 'onlineSavingsMsg'
Learn More
[1-(6-Methylpyrazin-2-yl)piperid-4-yl]methanol, 97%, Thermo Scientific™ Product Code.: 10461514

[1-(6-Methylpyrazin-2-yl)piperid-4-yl]methanol, 97%, Thermo Scientific™

Product Code. 10461514
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10461514

Brand: Thermo Scientific Maybridge CC58909DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 886851-59-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H17N3O
MDL Number MFCD09025896
Quantity 1 g
Synonym 1-6-methylpyrazin-2-yl piperid-4-yl methanol, 1-6-methylpyrazin-2-yl piperidin-4-yl methanol, 1-6-methylpyrazin-2-yl-4-piperidyl methan-1-ol
InChI Key KGGNRLCKRDFLQP-UHFFFAOYSA-N
SMILES CC1=CN=CC(=N1)N2CCC(CC2)CO
IUPAC Name [1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanol
Molecular Weight (g/mol) 207.277
PubChem CID 18525898
Formula Weight 207.28
Percent Purity 97%
Chemical Name or Material [1-(6-Methylpyrazin-2-yl)piperid-4-yl]methanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
[1-(6-Methylpyrazin-2-yl)piperid-4-yl]methanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.