missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-4-(n-octyloxy)benzene, 98%, Thermo Scientific Chemicals Product Code.: 11420953

1-Bromo-4-(n-octyloxy)benzene, 98%, Thermo Scientific Chemicals

Product Code. 11420953
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11420953

Brand: Thermo Scientific Alfa Aesar A13334.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96693-05-9
Molecular Formula C14H21BrO
Molecular Weight (g/mol) 285.225
MDL Number MFCD00043445
InChI Key UVBFFPZGOOKWNR-UHFFFAOYSA-N
Synonym p-bromophenyl octyl ether, 1-bromo-4-octyloxy benzene, 1-bromo-4-octyloxybenzene, 4-n-octyloxybromobenzene, 1-bromo-4-n-octyloxy benzene, 4-bromo-1-octyloxybenzene, 4-octyloxybromobenzene, 4-bromo-octyloxybenzene, 4-bromophenyloctyl ether, 1-4-bromophenoxy octaneShow MoreShow Less
PubChem CID 145298
IUPAC Name 1-bromo-4-octoxybenzene
SMILES CCCCCCCCOC1=CC=C(C=C1)Br

Specifications

Boiling Point 135°C to 137°C (0.5 mmHg)
Refractive Index 1.518
Quantity 1 g
Formula Weight 285.23
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-(n-octyloxy)benzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10223784 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13334.03,A13334.14
Lot NumberF9015C Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13334.03,A13334.14
Lot Number10213320 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13334.03,A13334.14
Lot Number10138221 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13334.03,A13334.14
Lot Number10223784 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13334.03,A13334.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-4-(n-octyloxy)benzene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.