missing translation for 'onlineSavingsMsg'
Learn More
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals Product Code.: 11405533

1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11405533
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11405533

Brand: Thermo Scientific Alfa Aesar A13459.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-87-6
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.452
MDL Number MFCD00001053
InChI Key GWQSENYKCGJTRI-UHFFFAOYSA-N
Synonym 4-chloroiodobenzene, p-chloroiodobenzene, 4-iodochlorobenzene, p-iodochlorobenzene, benzene, 1-chloro-4-iodo, 1-chloro-4-iodo-benzene, p-chlorjodbenzol, 4-chloro iodobenzene, 4-chloro-iodobenzene, 4-iodo-chlorobenzeneShow MoreShow Less
PubChem CID 12510
IUPAC Name 1-chloro-4-iodobenzene
SMILES C1=CC(=CC=C1Cl)I

Specifications

Melting Point 52°C to 56°C
Boiling Point 226°C to 227°C
Flash Point 108°C (226°F)
Quantity 25 g
Beilstein 1634414
Sensitivity Light sensitive
Formula Weight 238.46
Percent Purity 99%
Chemical Name or Material 1-Chloro-4-iodobenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248334 Certificate TypeCertificate of Analysis Date19/10/2023 Catalog NumberA13459.22,A13459.14,A13459.36
Lot Number10162612 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13459.22,A13459.14,A13459.36
Lot Number10110592 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13459.22,A13459.14,A13459.36
Lot Number10226341 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13459.22,A13459.14,A13459.36
Lot Number10149927 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13459.22,A13459.14,A13459.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.