missing translation for 'onlineSavingsMsg'
Learn More
1-Cyclohexene-1-carboxylic acid, 96% Product Code.: 10217460

1-Cyclohexene-1-carboxylic acid, 96%

Product Code. 10217460 Shop All Thermo Scientific Chemicals Products
2.5 g, Glass bottle, 2.50g
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.50g
This item is not returnable. View return policy

Product Code. 10217460

Brand: Thermo Scientific Acros 291750025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 636-82-8 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 636-82-8
CAS Min % 95.0
CAS Max % 100.0
Melting Point 35.0°C to 39.0°C
Color White to Yellow
Boiling Point 137.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H10O2
Refractive Index 1.5
MDL Number MFCD00001545
Quantity 2.5 g
Synonym 1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid
InChI Key NMEZJSDUZQOPFE-UHFFFAOYSA-N
SMILES C1CCC(=CC1)C(=O)O
IUPAC Name cyclohexene-1-carboxylic acid
Molecular Weight (g/mol) 126.16
PubChem CID 69470
ChEBI CHEBI:29565
Formula Weight 126.16
Percent Purity 96%
Physical Form Crystalline Mass or Liquid
Chemical Name or Material 1-Cyclohexene-1-carboxylic acid, 96%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0388190 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code291750025
Lot NumberA0388190 Certificate TypeCertificate of Analysis Date01/09/2023 Product Code291750025
Lot NumberA0376225 Certificate TypeCertificate of Analysis Date14/12/2022 Product Code291750025
compliance-icons
Product Identifier
  • 1-Cyclohexene-1-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Cyclohexene-1-carboxylic acid, 96% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.