missing translation for 'onlineSavingsMsg'
Learn More
1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals Product Code.: 11410388

1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals

Product Code. 11410388
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11410388

Brand: Thermo Scientific Alfa Aesar B24548.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-91-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00003199
InChI Key ZFPGARUNNKGOBB-UHFFFAOYSA-N
Synonym 1-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, n-ethylpyrrolidinone, ethyl pyrrolidone, unii-h0229sx1cw, n-ethyl-, a-pyrrolidone, n-ethyl-2-pyrrolidinoneShow MoreShow Less
PubChem CID 17595
IUPAC Name 1-ethylpyrrolidin-2-one
SMILES CCN1CCCC1=O

Specifications

Density 0.992
Boiling Point 96°C to 98°C (20 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.465
Quantity 100 g
Formula Weight 113.16
Percent Purity 98%
Chemical Name or Material 1-Ethyl-2-pyrrolidinone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10127055 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberB24548.22,B24548.14,B24548.36
Lot Number10127055 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24548.22,B24548.14,B24548.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.