Learn More
1-Heptanesulfonic acid sodium salt monohydrate, ∽98%, MP Biomedicals™
41.65€ - 228.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17861361
|
MP Biomedicals
0215123905 |
5 g |
41.65€
5g |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
17861371
|
MP Biomedicals
0215123925 |
25 g |
228.00€
25g |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||