missing translation for 'onlineSavingsMsg'
Learn More
1-Phenyloctane, 99%, Thermo Scientific Chemicals Product Code.: 10561374

1-Phenyloctane, 99%, Thermo Scientific Chemicals

Product Code. 10561374
100 mL, Glass bottle
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10561374

Brand: Thermo Scientific Acros 150991000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2189-60-8
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00009564
InChI Key CDKDZKXSXLNROY-UHFFFAOYSA-N
Synonym n-octylbenzene, 1-phenyloctane, benzene, octyl, phenyloctane, octane, 1-phenyl, octyl-benzene, 1-octylbenzene, octyl benzene, 4-octylbenzene, acmc-1cplgShow MoreShow Less
PubChem CID 16607
IUPAC Name octylbenzene
SMILES CCCCCCCCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -36°C
Color Colorless
Density 0.8500g/mL
Boiling Point 261°C to 263°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)7CH3
Refractive Index 1.4830 to 1.485
Quantity 100 mL
Beilstein 05, 453
Specific Gravity 0.85
Formula Weight 190.33
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Phenyloctane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468752 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number150990250,150991000
Lot NumberA0454924 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number150990250,150991000
Lot NumberA0450225 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number150990250,150991000
Lot NumberA0410155 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number150990250,150991000
Lot NumberA0438270 Certificate TypeCertificate of Analysis Date25/01/2022 Catalog Number150990250,150991000
Product Identifier
  • 1-Phenyloctane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Phenyloctane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.