Learn More
Thermo Scientific Alfa Aesar 100GR Ampicillin trihydrate
Brand: Thermo Scientific Alfa Aesar J66514.22
100GR Ampicillin trihydrate
Description

Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Ampicillin trihydrate
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Ampicillin trihydrate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P20K035 | Certificate of Analysis | 02/04/2024 | J66514.14,J66514.36,J66514.22 |
P20K020 | Certificate of Analysis | 02/04/2024 | J66514.14,J66514.36,J66514.22 |
U10J019 | Certificate of Analysis | 24/10/2023 | J66514.14,J66514.36,J66514.22 |
S08J049 | Certificate of Analysis | 24/10/2023 | J66514.14,J66514.36,J66514.22 |
Y18H011 | Certificate of Analysis | 17/11/2022 | J66514.14,J66514.36,J66514.22 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.