Learn More
Fisher Chemical 10KG POTASSIUM CHLORIDE 99.5 % FOR ANALYSIS
Brand:  Fisher Chemical P/4280/68
884.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
10KG POTASSIUM CHLORIDE 99.5 % FOR ANALYSIS
Product Code. 10376190
  • 890.00€ / 10kg
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2529617 Certificate TypeCertificate of Analysis Date13/03/2025 Catalog NumberP/4280/53,P/4280/68,P/4280/70,P/4280/60
Product Title
Fisher Chemical 10KG POTASSIUM CHLORIDE 99.5 % FOR ANALYSIS >