Learn More
Fisher Chemical 10KG SUCCINIC ACID 99.5+ %
Brand:  Fisher Chemical S/8522/68
539.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
10KG SUCCINIC ACID 99.5+ %
Product Code. 10628333
  • 540.00€ / 10kg
Explore more special offers
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Succinic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisher Chemical 10KG SUCCINIC ACID 99.5+ % >