Learn More
Fisher Chemical 10KG SUCCINIC ACID 99.5+ %
Brand: Fisher Chemical S/8522/68
539.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
10KG SUCCINIC ACID 99.5+ %
This item is not returnable.
View return policy
Description
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Succinic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Product Title
Fisher Chemical 10KG SUCCINIC ACID 99.5+ % >