missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products 1422A DuPont™ Tyvek™ Autoclave Sterilization Wrapper
GREENER_CHOICE

Autoclavable Wrapper provides an excellent bacterial and microbial barrier for superior performance post steam sterilization.

Brand:  Keystone Cleanroom Products T2ASHEET1818

 View more versions of this product

Product Code. 17748537

  • 1350.00€ / Pack of 500
Only null left
Estimated Shipment: 24-07-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Tyvek™ Autoclavable Wrappers are breathable while providing an excellent bacterial and microbial barrier for superior performance post steam sterilization.

  • Lint-free (Helmke Drum) means no airborne particulates are generated during its use as sterilization packaging
  • Clean and consistently cut using an automated CAD system per ISO 8 Requirements
  • Packaged in an ISO 6 Controlled Cleanroom
  • Designed for Steam Sterilization
  • Hydrophobic
  • Resists tearing and punctures
Specifications

Specifications

Autoclavable
None
Single-use
Pharmaceutical Production Equipment
Non-sterile
Yes
Steam, EtO, Gamma, VHP
<125°C
18 in.
18 in.
Sterilization Wrap
COC
White
Steam Sterilization
Tyvek
5.3 mm
500
<250°F
18 x 18 in.
45.7 cm
45.7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Keystone Cleanroom Products 1422A DuPont™ Tyvek™ Autoclave Sterilization Wrapper > Dimensions (L x W): 18 x 18 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.