missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 1536-well HiBase Flat Bottom Microplate
Used for reduced sample consumption and assay miniaturization.
Brand: Greiner Bio-One 782061
This item is not returnable.
View return policy
Description
- Sterile
- High Base
- High binding
Specifications
Standard | |
Not Autoclavable | |
Clear | |
1536 | |
Microlon 600 | |
Flat Bottom (F-well) | |
60 | |
-20°C to +60°C |
32 x 48 | |
High Binding | |
Polystyrene | |
Sterile | |
6 to 12 μL | |
1800 x g (swinging-bucket rotor) | |
15 |