Learn More
Greiner Bio-One 1536 Well HiBase Microplates
Designed with a rounded square profile for flexible working volume combined with reduced wicking and bubbling. Greiner Bio-One™ 1536 Well HiBase Microplates are ideally suited for top reading instruments, fluorescence measurements and, due to hydrophobic nature of surface, non polar proteins and peptides.
Brand: Greiner Bio-One 782076
This item is not returnable.
View return policy
Description
- Clear polystyrene construction for optimum visibility
- Can be traced to production through defined LOT number system
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Square wells with rounded corners combines advantages of square well (i.e. flexible working volume of 3μ to 10μ with the advantages of a round well (reduced wicking and bubbling)
Specifications
DIN ISO 9001 | |
Standard | |
32 x 48 | |
Polystyrene | |
3 to 10 μL | |
FLUOTRAC 200 | |
Flat Bottom (F-well) |
Free of detectable DNase, RNase, human DNA, non-pyrogenic | |
Without Lid | |
Black | |
1536 | |
Non-sterile | |
12.6 μL | |
60/Cs. |