Learn More
Greiner Bio-One 1536 Well HiBase Microplates
Designed with a rounded square profile for flexible working volume combined with reduced wicking and bubbling. Greiner Bio-One™ 1536 Well HiBase Microplates are ideally suited for top reading instruments, fluorescence measurements and, due to hydrophobic nature of surface, non polar proteins and peptides.
Brand:  Greiner Bio-One 782076
Product Code. 11719512
  • 4460.00€ / Pack of 60
Estimated Shipment: 26-11-2024
Description
  • Clear polystyrene construction for optimum visibility
  • Can be traced to production through defined LOT number system
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Square wells with rounded corners combines advantages of square well (i.e. flexible working volume of 3μ to 10μ with the advantages of a round well (reduced wicking and bubbling) 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN ISO 9001
Standard
32 x 48
Polystyrene
3 to 10 μL
FLUOTRAC 200
Flat Bottom (F-well)
Free of detectable DNase, RNase, human DNA, non-pyrogenic
Without Lid
Black
1536
Non-sterile
12.6 μL
60/Cs.